Assay Details
| Gene Symbol: AT5G35320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122923.3 | NP_198382.1 | 2 - 3 | 442 | 138 | |
| GenBank mRNA | AY074341.1 | - | 2 - 3 | 342 | 138 | |
| AY117159.1 | - | 2 - 3 | 295 | 138 | ||
More Information
Set Membership: |
|

AT5G35320
Genomic Map