Assay Details
| Gene Symbol: CAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036891.2 | NP_001031968.1 | 4 - 5 | 771 | 94 | |
| NM_001203493.1 | NP_001190422.1 | 4 - 5 | 823 | 94 | ||
| NM_122927.4 | NP_198386.1 | 4 - 5 | 825 | 94 | ||
| GenBank mRNA | AY085968.1 | - | 4 - 5 | 768 | 94 | |
| AY142630.1 | - | 4 - 5 | 629 | 94 | ||
| U90879.1 | - | 4 - 5 | 747 | 94 | ||
More Information
Set Membership: |
|

CAC2
Genomic Map