Assay Details
| Gene Symbol: PAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122950.4 | NP_198409.1 | 2 - 3 | 298 | 75 | |
| GenBank mRNA | AK228466.1 | - | 2 - 3 | 230 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAA1| Gene Symbol: PAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122950.4 | NP_198409.1 | 2 - 3 | 298 | 75 | |
| GenBank mRNA | AK228466.1 | - | 2 - 3 | 230 | 75 | |
Set Membership: |
|