Assay Details
| Gene Symbol: PAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122950.4 | NP_198409.1 | 4 - 5 | 472 | 107 | |
| GenBank mRNA | AK228466.1 | - | 4 - 5 | 404 | 107 | |
More Information
Set Membership: |
|

PAA1
Genomic Map