Assay Details
| Gene Symbol: GS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036892.1 | NP_001031969.1 | 10 - 11 | 1269 | 76 | |
| NM_001085170.1 | NP_001078639.1 | 10 - 11 | 1258 | 76 | ||
| NM_122954.4 | NP_198413.1 | 10 - 11 | 1483 | 76 | ||
| GenBank mRNA | AF428319.1 | - | 10 - 11 | 1263 | 76 | |
| AF428461.1 | - | 10 - 11 | 1263 | 76 | ||
| AK317110.1 | - | 10 - 11 | 1270 | 76 | ||
| AY081252.1 | - | 10 - 11 | 1217 | 76 | ||
| AY088222.1 | - | 10 - 11 | 1296 | 76 | ||
| AY091114.1 | - | 10 - 11 | 1265 | 76 | ||
| AY122977.1 | - | 9 - 10 | 1159 | 76 | ||
| S69727.1 | - | 10 - 11 | 1230 | 76 | ||
More Information
Set Membership: |
|

GS2
Genomic Map