Assay Details
| Gene Symbol: GS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036892.1 | NP_001031969.1 | 9 - 10 | 1203 | 65 | |
| NM_001085170.1 | NP_001078639.1 | 9 - 10 | 1192 | 65 | ||
| NM_122954.4 | NP_198413.1 | 9 - 10 | 1417 | 65 | ||
| GenBank mRNA | AF428319.1 | - | 9 - 10 | 1197 | 65 | |
| AF428461.1 | - | 9 - 10 | 1197 | 65 | ||
| AK317110.1 | - | 9 - 10 | 1204 | 65 | ||
| AY081252.1 | - | 9 - 10 | 1151 | 65 | ||
| AY088222.1 | - | 9 - 10 | 1230 | 65 | ||
| AY091114.1 | - | 9 - 10 | 1199 | 65 | ||
| AY122977.1 | - | 8 - 9 | 1093 | 65 | ||
| S69727.1 | - | 9 - 10 | 1164 | 65 | ||
More Information
Set Membership: |
|

GS2
Genomic Map