Assay Details
| Gene Symbol: G6PD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122970.6 | NP_198428.1 | 4 - 5 | 921 | 110 | |
| GenBank mRNA | AY086213.1 | - | 4 - 5 | 820 | 110 | |
| AY099561.1 | - | 4 - 5 | 823 | 110 | ||
| BT002133.1 | - | 4 - 5 | 777 | 110 | ||
More Information
Set Membership: |
|

G6PD1
Genomic Map