Assay Details
| Gene Symbol: G6PD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_122970.6 | NP_198428.1 | 5 - 6 | 1068 | 102 | |
| GenBank mRNA | AY086213.1 | - | 5 - 6 | 967 | 102 | |
| AY099561.1 | - | 5 - 6 | 970 | 102 | ||
| BT002133.1 | - | 5 - 6 | 924 | 102 | ||
More Information
Set Membership: |
|

G6PD1
Genomic Map