Assay Details
| Gene Symbol: BIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344271.1 | NP_001330505.1 | 2 - 3 | 352 | 127 | |
| NM_123247.5 | NP_198702.2 | 2 - 3 | 407 | 127 | ||
| GenBank mRNA | AJ630499.1 | - | 2 - 3 | 225 | 127 | |
| AY568671.1 | - | 2 - 3 | 225 | 127 | ||
| BT033131.1 | - | 2 - 3 | 225 | 127 | ||
More Information
Set Membership: |
|

BIM3
Genomic Map