Assay Details
| Gene Symbol: cdc2cAt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123304.4 | NP_198758.2 | 7 - 8 | 2091 | 78 | |
| GenBank mRNA | BT046183.1 | - | 7 - 8 | 1790 | 78 | |
More Information
Set Membership: |
|

cdc2cAt
Genomic Map