Assay Details
| Gene Symbol: NAC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123323.3 | NP_198777.1 | 1 - 2 | 456 | 103 | |
| GenBank mRNA | AY056327.1 | - | 1 - 2 | 318 | 103 | |
| AY091191.1 | - | 1 - 2 | 195 | 103 | ||
| AY174163.1 | - | 1 - 2 | 195 | 103 | ||
More Information
Set Membership: |
|

NAC6
Genomic Map