Assay Details
| Gene Symbol: AT5G39910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123354.1 | NP_198807.1 | 6 - 7 | 735 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G39910| Gene Symbol: AT5G39910 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123354.1 | NP_198807.1 | 6 - 7 | 735 | 140 | |
Set Membership: |
|