Assay Details
| Gene Symbol: SWEET8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036909.1 | NP_001031986.1 | 2 - 3 | 170 | 136 | |
| NM_123390.3 | NP_568579.1 | 2 - 3 | 249 | 136 | ||
| GenBank mRNA | AK118347.1 | - | 2 - 3 | 165 | 136 | |
| AY084840.1 | - | 2 - 3 | 169 | 136 | ||
| BT005477.1 | - | 2 - 3 | 85 | 136 | ||
More Information
Set Membership: |
|

SWEET8
Genomic Map