Assay Details
| Gene Symbol: ERA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344319.1 | NP_001331518.1 | 3 - 4 | 429 | 132 | |
| NM_123392.2 | NP_198844.1 | 3 - 4 | 477 | 132 | ||
| GenBank mRNA | AF214106.1 | - | 3 - 4 | 429 | 132 | |
| BT030457.1 | - | 3 - 4 | 429 | 132 | ||
More Information
Set Membership: |
|

ERA1
Genomic Map