Assay Details
| Gene Symbol: ERA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123392.2 | NP_198844.1 | 5 - 6 | 591 | 77 | |
| GenBank mRNA | AF214106.1 | - | 5 - 6 | 543 | 77 | |
| BT030457.1 | - | 5 - 6 | 543 | 77 | ||
More Information
Set Membership: |
|

ERA1
Genomic Map