Assay Details
| Gene Symbol: ERA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344319.1 | NP_001331518.1 | 6 - 7 | 748 | 76 | |
| NM_123392.2 | NP_198844.1 | 7 - 8 | 700 | 76 | ||
| GenBank mRNA | AF214106.1 | - | 7 - 8 | 652 | 76 | |
| BT030457.1 | - | 7 - 8 | 652 | 76 | ||
More Information
Set Membership: |
|

ERA1
Genomic Map