Assay Details
| Gene Symbol: AT5G40290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123393.1 | NP_198845.1 | 10 - 11 | 676 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G40290| Gene Symbol: AT5G40290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123393.1 | NP_198845.1 | 10 - 11 | 676 | 112 | |
Set Membership: |
|