Assay Details
| Gene Symbol: AT5G40300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123394.4 | NP_198846.1 | 1 - 2 | 628 | 138 | |
| GenBank mRNA | BT010719.1 | - | 1 - 2 | 574 | 138 | |
| BT011792.1 | - | 1 - 2 | 484 | 138 | ||
More Information
Set Membership: |
|

AT5G40300
Genomic Map