Assay Details
| Gene Symbol: AT5G40300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123394.4 | NP_198846.1 | 2 - 3 | 758 | 76 | |
| GenBank mRNA | BT010719.1 | - | 2 - 3 | 704 | 76 | |
| BT011792.1 | - | 2 - 3 | 614 | 76 | ||
More Information
Set Membership: |
|

AT5G40300
Genomic Map