Assay Details
| Gene Symbol: MYB115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123400.2 | NP_568581.1 | 2 - 3 | 1051 | 95 | |
| GenBank mRNA | DQ447019.1 | - | 2 - 3 | 623 | 95 | |
More Information
Set Membership: |
|

MYB115
Genomic Map