Assay Details
| Gene Symbol: SIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123403.4 | NP_198855.1 | 1 - 2 | 1146 | 94 | |
| GenBank mRNA | AY062781.1 | - | 1 - 2 | 1006 | 94 | |
| AY081645.1 | - | 1 - 2 | 875 | 94 | ||
More Information
Set Membership: |
|

SIP1
Genomic Map