Assay Details
| Gene Symbol: TOM20-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344374.1 | NP_001330863.1 | 3 - 4 | 404 | 126 | |
| NM_123458.5 | NP_198909.1 | 3 - 4 | 404 | 126 | ||
| GenBank mRNA | AF462861.1 | - | 3 - 4 | 401 | 126 | |
| AK117822.1 | - | 3 - 4 | 389 | 126 | ||
| AY141998.1 | - | 3 - 4 | 338 | 126 | ||
More Information
Set Membership: |
|

TOM20-4
Genomic Map