Assay Details
| Gene Symbol: TOM20-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123458.5 | NP_198909.1 | 5 - 6 | 513 | 79 | |
| GenBank mRNA | AF462861.1 | - | 5 - 6 | 510 | 79 | |
| AK117822.1 | - | 5 - 6 | 498 | 79 | ||
| AY141998.1 | - | 5 - 6 | 447 | 79 | ||
More Information
Set Membership: |
|

TOM20-4
Genomic Map