Assay Details
| Gene Symbol: DRB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123472.3 | NP_198923.2 | 2 - 3 | 438 | 83 | |
| GenBank mRNA | AK117808.1 | - | 2 - 3 | 396 | 83 | |
| BT009653.1 | - | 1 - 2 | 248 | 83 | ||
More Information
Set Membership: |
|

DRB5
Genomic Map