Assay Details
| Gene Symbol: DRB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123472.3 | NP_198923.2 | 3 - 4 | 657 | 76 | |
| GenBank mRNA | AK117808.1 | - | 3 - 4 | 615 | 76 | |
| BT009653.1 | - | 2 - 3 | 467 | 76 | ||
More Information
Set Membership: |
|

DRB5
Genomic Map