Assay Details
| Gene Symbol: UVH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344388.1 | NP_001330264.1 | 7 - 8 | 2957 | 83 | |
| NM_123480.4 | NP_198931.1 | 8 - 9 | 3161 | 83 | ||
| GenBank mRNA | AF089003.1 | - | 8 - 9 | 2663 | 83 | |
| AF160500.1 | - | 8 - 9 | 2757 | 83 | ||
| AY140076.1 | - | 8 - 9 | 2695 | 83 | ||
More Information
Set Membership: |
|

UVH1
Genomic Map