Assay Details
| Gene Symbol: AT5G41310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123496.2 | NP_198947.1 | 15 - 16 | 2092 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G41310| Gene Symbol: AT5G41310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123496.2 | NP_198947.1 | 15 - 16 | 2092 | 148 | |
Set Membership: |
|