Assay Details
| Gene Symbol: XPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123502.3 | NP_568592.1 | 6 - 7 | 792 | 65 | |
| GenBank mRNA | AY039547.1 | - | 6 - 7 | 763 | 65 | |
| AY113008.1 | - | 5 - 6 | 574 | 65 | ||
| U29168.2 | - | 6 - 7 | 792 | 65 | ||
More Information
Set Membership: |
|

XPB1
Genomic Map