Assay Details
| Gene Symbol: RPS10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123516.4 | NP_198967.1 | 2 - 3 | 161 | 107 | |
| GenBank mRNA | AF375440.1 | - | 2 - 3 | 130 | 107 | |
| AY037240.1 | - | 2 - 3 | 130 | 107 | ||
| AY060539.1 | - | 1 - 2 | 52 | 107 | ||
More Information
Set Membership: |
|

RPS10B
Genomic Map