Assay Details
| Gene Symbol: AT5G41800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123545.4 | NP_568597.1 | 4 - 5 | 780 | 115 | |
| GenBank mRNA | AY099685.1 | - | 4 - 5 | 612 | 115 | |
| AY128864.1 | - | 4 - 5 | 601 | 115 | ||
More Information
Set Membership: |
|

AT5G41800
Genomic Map