Assay Details
| Gene Symbol: AT5G41870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123552.2 | NP_199002.1 | 1 - 2 | 432 | 60 | |
| GenBank mRNA | BT015129.1 | - | 1 - 2 | 426 | 60 | |
More Information
Set Membership: |
|

AT5G41870
Genomic Map