Assay Details
| Gene Symbol: MED10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344425.1 | NP_001331138.1 | 1 - 2 | 476 | 140 | |
| NM_001344426.1 | NP_001331137.1 | 2 - 3 | 263 | 140 | ||
| NM_123556.3 | NP_199006.1 | 2 - 3 | 291 | 140 | ||
| GenBank mRNA | BT002847.1 | - | 2 - 3 | 270 | 140 | |
| BT004431.1 | - | 1 - 2 | 213 | 140 | ||
More Information
Set Membership: |
|

MED10A
Genomic Map