Assay Details
| Gene Symbol: ABIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344431.1 | NP_001330354.1 | 2 - 3 | 310 | 69 | |
| NM_001344432.1 | NP_001330353.1 | 2 - 3 | 279 | 69 | ||
| NM_123568.5 | NP_199018.1 | 2 - 3 | 391 | 69 | ||
| GenBank mRNA | AK175528.1 | - | 2 - 3 | 223 | 69 | |
| AK175563.1 | - | 2 - 3 | 192 | 69 | ||
| AK176699.1 | - | 2 - 3 | 279 | 69 | ||
| AK176912.1 | - | 2 - 3 | 187 | 69 | ||
| AY817015.1 | - | 2 - 3 | 183 | 69 | ||
| BT024503.1 | - | 2 - 3 | 183 | 69 | ||
More Information
Set Membership: |
|

ABIL4
Genomic Map