Assay Details
| Gene Symbol: ABIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344431.1 | NP_001330354.1 | 5 - 6 | 612 | 100 | |
| NM_001344432.1 | NP_001330353.1 | 5 - 6 | 581 | 100 | ||
| NM_001344433.1 | NP_001330352.1 | 3 - 4 | 393 | 100 | ||
| NM_123568.5 | NP_199018.1 | 5 - 6 | 693 | 100 | ||
| GenBank mRNA | AK175528.1 | - | 5 - 6 | 525 | 100 | |
| AK175563.1 | - | 5 - 6 | 494 | 100 | ||
| AK176699.1 | - | 5 - 6 | 581 | 100 | ||
| AK176912.1 | - | 5 - 6 | 489 | 100 | ||
| AY817015.1 | - | 5 - 6 | 485 | 100 | ||
| BT024503.1 | - | 5 - 6 | 485 | 100 | ||
More Information
Set Membership: |
|

ABIL4
Genomic Map