Assay Details
| Gene Symbol: ABIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344431.1 | NP_001330354.1 | 7 - 8 | 725 | 88 | |
| NM_001344432.1 | NP_001330353.1 | 6 - 7 | 764 | 88 | ||
| NM_001344433.1 | NP_001330352.1 | 5 - 6 | 506 | 88 | ||
| NM_123568.5 | NP_199018.1 | 7 - 8 | 806 | 88 | ||
| GenBank mRNA | AK175528.1 | - | 7 - 8 | 638 | 88 | |
| AK175563.1 | - | 7 - 8 | 607 | 88 | ||
| AK176699.1 | - | 7 - 8 | 694 | 88 | ||
| AK176912.1 | - | 7 - 8 | 602 | 88 | ||
| AY817015.1 | - | 7 - 8 | 598 | 88 | ||
| BT024503.1 | - | 7 - 8 | 598 | 88 | ||
More Information
Set Membership: |
|

ABIL4
Genomic Map