Assay Details
| Gene Symbol: AT5G42190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123584.5 | NP_568603.1 | 1 - 2 | 570 | 75 | |
| GenBank mRNA | AF059295.1 | - | 1 - 2 | 364 | 75 | |
| AK227773.1 | - | 1 - 2 | 393 | 75 | ||
| BT004798.1 | - | 1 - 2 | 345 | 75 | ||
More Information
Set Membership: |
|

AT5G42190
Genomic Map