Assay Details
| Gene Symbol: AT5G42420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085220.1 | NP_001078689.1 | 1 - 2 | 293 | 89 | |
| NM_123607.5 | NP_199057.1 | 1 - 2 | 296 | 89 | ||
| GenBank mRNA | AK228744.1 | - | 1 - 2 | 287 | 89 | |
| BT025259.1 | - | 1 - 2 | 132 | 89 | ||
| KJ667159.1 | - | 1 - 2 | 132 | 89 | ||
| KP872770.1 | - | 1 - 2 | 132 | 89 | ||
More Information
Set Membership: |
|

AT5G42420
Genomic Map