Assay Details
| Gene Symbol: AT5G42420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123607.5 | NP_199057.1 | 2 - 3 | 480 | 94 | |
| GenBank mRNA | AK228744.1 | - | 2 - 3 | 471 | 94 | |
| BT025259.1 | - | 2 - 3 | 316 | 94 | ||
| KJ667159.1 | - | 2 - 3 | 316 | 94 | ||
| KP872770.1 | - | 2 - 3 | 316 | 94 | ||
More Information
Set Membership: |
|

AT5G42420
Genomic Map