Assay Details
| Gene Symbol: AT5G42470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123612.4 | NP_199062.2 | 12 - 13 | 1301 | 107 | |
| GenBank mRNA | BT015735.1 | - | 12 - 13 | 1128 | 107 | |
| BT020175.1 | - | 12 - 13 | 1085 | 107 | ||
More Information
Set Membership: |
|

AT5G42470
Genomic Map