Assay Details
| Gene Symbol: CYP71A16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123623.5 | NP_199073.1 | 2 - 3 | 1035 | 85 | |
| GenBank mRNA | BT026460.1 | - | 2 - 3 | 875 | 85 | |
More Information
Set Membership: |
|

CYP71A16
Genomic Map