Assay Details
| Gene Symbol: HB27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123643.3 | NP_199092.1 | 1 | 728 | 74 | |
| GenBank mRNA | BT002832.1 | - | 1 | 670 | 74 | |
| BT004379.1 | - | 1 | 605 | 74 | ||
| LN852659.1 | - | 1 | 605 | 74 | ||
More Information
Set Membership: |
|

HB27
Genomic Map