Assay Details
| Gene Symbol: AT5G42830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123648.3 | NP_199097.1 | 1 - 2 | 549 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G42830| Gene Symbol: AT5G42830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123648.3 | NP_199097.1 | 1 - 2 | 549 | 81 | |
Set Membership: |
|