Assay Details
| Gene Symbol: AT5G42840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203527.2 | NP_001190456.1 | 2 | 1924 | 118 | |
| NM_123649.2 | NP_199098.1 | 1 | 2039 | 118 | ||
More Information
Set Membership: |
|

AT5G42840
Genomic Map