Assay Details
| Gene Symbol: AT5G42860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123651.4 | NP_199100.1 | 1 - 2 | 597 | 80 | |
| GenBank mRNA | AK227557.1 | - | 1 - 2 | 451 | 80 | |
More Information
Set Membership: |
|

AT5G42860
Genomic Map