Assay Details
| Gene Symbol: AT5G42940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001344467.1 | NP_001330419.1 | 2 - 3 | 2000 | 65 | |
| NM_123659.4 | NP_199108.1 | 2 - 3 | 2024 | 65 | ||
| GenBank mRNA | AF462851.1 | - | 2 - 3 | 1813 | 65 | |
| AK319077.1 | - | 1 - 2 | 2082 | 65 | ||
| BT004548.1 | - | 1 - 2 | 1626 | 65 | ||
More Information
Set Membership: |
|

AT5G42940
Genomic Map