Assay Details
| Gene Symbol: AT5G42950 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123660.3 | NP_199109.1 | 3 - 4 | 1205 | 81 | |
| GenBank mRNA | LC130495.1 | - | 3 - 4 | 1120 | 81 | |
More Information
Set Membership: |
|

AT5G42950
Genomic Map