Assay Details
| Gene Symbol: COP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123663.3 | NP_199111.1 | 11 - 12 | 1228 | 86 | |
| GenBank mRNA | AF395060.1 | - | 11 - 12 | 1061 | 86 | |
| AK176360.1 | - | 11 - 12 | 1143 | 86 | ||
| BT025548.1 | - | 11 - 12 | 1061 | 86 | ||
More Information
Set Membership: |
|

COP8
Genomic Map