Assay Details
| Gene Symbol: COP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123663.3 | NP_199111.1 | 9 - 10 | 1031 | 81 | |
| GenBank mRNA | AF395060.1 | - | 9 - 10 | 864 | 81 | |
| AK176360.1 | - | 9 - 10 | 946 | 81 | ||
| BT025548.1 | - | 9 - 10 | 864 | 81 | ||
More Information
Set Membership: |
|

COP8
Genomic Map