Assay Details
| Gene Symbol: RPT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123667.4 | NP_199115.1 | 8 - 9 | 1208 | 74 | |
| GenBank mRNA | AF123393.1 | - | 8 - 9 | 1030 | 74 | |
| AF372945.1 | - | 8 - 9 | 1142 | 74 | ||
| AY078040.1 | - | 8 - 9 | 1022 | 74 | ||
More Information
Set Membership: |
|

RPT4A
Genomic Map