Assay Details
| Gene Symbol: AT5G43230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123689.3 | NP_199137.2 | 2 - 3 | 1780 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT5G43230| Gene Symbol: AT5G43230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_123689.3 | NP_199137.2 | 2 - 3 | 1780 | 79 | |
Set Membership: |
|